missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl phosphate, disodium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-4090121
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
207569-06-0, 2183-17-7
C10H7Na2O4P
268.12
MFCD00041007
QYURIFWAOPAPAJ-UHFFFAOYSA-L
1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
24212014
disodium;naphthalen-1-yl phosphate;hydrate
[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1
This item is not returnable. View return policy
207569-06-0, 2183-17-7
C10H7Na2O4P
268.12
MFCD00041007
QYURIFWAOPAPAJ-UHFFFAOYSA-L
1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
24212014
disodium;naphthalen-1-yl phosphate;hydrate
[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 286.13
Percent Purity 98%
Chemical Name or Material 1-Naphthyl phosphate, disodium salt hydrate
compliance-icons
Product Identifier
  • 1-Naphthyl phosphate, disodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'