Learn More
Alfa Aesar™ 1-Nitro-4-n-propylbenzene, 96%
Brand: Alfa Aesar™ L07730.14
Additional Details : CAS : 10342-59-3 Weight : 0.02500kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
10342-59-3 | |
165.192 | |
SXQBFCVVZIYXHV-UHFFFAOYSA-N | |
82559 | |
CCCC1=CC=C(C=C1)[N+](=O)[O-] |
C9H11NO2 | |
MFCD00051839 | |
1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol | |
1-nitro-4-propylbenzene |
Specifications
1-Nitro-4-n-propylbenzene | |
96°C to 100°C (2mmHg) | |
25g | |
C9H11NO2 | |
UN2810 | |
1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol | |
CCCC1=CC=C(C=C1)[N+](=O)[O-] | |
165.192 | |
165.19 |
1.537 | |
-14°C | |
10342-59-3 | |
MFCD00051839 | |
2043939 | |
SXQBFCVVZIYXHV-UHFFFAOYSA-N | |
1-nitro-4-propylbenzene | |
82559 | |
96% |