missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-4-n-propylbenzene, 96%, Thermo Scientific Chemicals

Catalog Number p-7044678
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
10342-59-3
C9H11NO2
165.19
MFCD00051839
SXQBFCVVZIYXHV-UHFFFAOYSA-N
1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol
82559
1-nitro-4-propylbenzene
CCCC1=CC=C(C=C1)[N+]([O-])=O
This item is not returnable. View return policy
10342-59-3
C9H11NO2
165.19
MFCD00051839
SXQBFCVVZIYXHV-UHFFFAOYSA-N
1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol
82559
1-nitro-4-propylbenzene
CCCC1=CC=C(C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -14°C
Boiling Point 96°C to 100°C (2 mmHg)
Refractive Index 1.537
Quantity 1 g
UN Number UN2810
Beilstein 2043939
Formula Weight 165.19
Percent Purity 96%
Chemical Name or Material 1-Nitro-4-n-propylbenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'