missing translation for 'onlineSavingsMsg'
Learn More

1-O-Methylsulfonyl 3-O-Trityl-(S,S)-Chloramphenicol, TRC

Product Code. 30474566
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
This item is not returnable. View return policy

Product Code. 30474566

Brand: Toronto Research Chemicals TRCM32736025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-O-Methylsulfonyl 3-O-Trityl-(S,S)-Chloramphenicol
Analyte or Component Names 1-O-Methylsulfonyl 3-O-Trityl-(S,S)-Chloramphenicol
Shipping Condition Room Temperature
Molecular Formula C31 H28 Cl2 N2 O7 S
Synonym (1S,2S)-2-(2,2-Dichloroacetamido)-1-(4-nitrophenyl)-3-(trityloxy)propyl Methanesulfonate
InChI Formula InChI=1S/C31H28Cl2N2O7S/c1-43(39,40)42-28(22-17-19-26(20-18-22)35(37)38)27(34-30(36)29(32)33)21-41-31(23-11-5-2-6-12-23,24-13-7-3-8-14-24)25-15-9-4-10-16-25/h2-20,27-29H,21H2,1H3,(H,34,36)/t27-,28-/m0/s1
Solution Type Neat
SMILES CS(=O)(=O)O[C@H]([C@H](COC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)C(Cl)Cl)c4ccc(cc4)[N+](=O)[O-]
IUPAC Name [(1S,2S)-2-[(2,2-dichloroacetyl)amino]-1-(4-nitrophenyl)-3-trityloxypropyl] methanesulfonate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 643.534
Quantity 25 mg
Formula Weight 642.099
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.