missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-O-Methylsulfonyl 3-O-Trityl-(S,S)-Chloramphenicol, TRC
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
Specifications
Specifications
| Chemical Name or Material | 1-O-Methylsulfonyl 3-O-Trityl-(S,S)-Chloramphenicol |
| Analyte or Component Names | 1-O-Methylsulfonyl 3-O-Trityl-(S,S)-Chloramphenicol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C31 H28 Cl2 N2 O7 S |
| Synonym | (1S,2S)-2-(2,2-Dichloroacetamido)-1-(4-nitrophenyl)-3-(trityloxy)propyl Methanesulfonate |
| InChI Formula | InChI=1S/C31H28Cl2N2O7S/c1-43(39,40)42-28(22-17-19-26(20-18-22)35(37)38)27(34-30(36)29(32)33)21-41-31(23-11-5-2-6-12-23,24-13-7-3-8-14-24)25-15-9-4-10-16-25/h2-20,27-29H,21H2,1H3,(H,34,36)/t27-,28-/m0/s1 |
| Solution Type | Neat |
| SMILES | CS(=O)(=O)O[C@H]([C@H](COC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)C(Cl)Cl)c4ccc(cc4)[N+](=O)[O-] |
| IUPAC Name | [(1S,2S)-2-[(2,2-dichloroacetyl)amino]-1-(4-nitrophenyl)-3-trityloxypropyl] methanesulfonate |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction