Learn More
1-Octadecene, tech. 90%, Thermo Scientific Chemicals
Description
1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Octadecene is an inexpensive solvent having high boiling point. Due to this property, it replaces the heat transfer fluids such as dowthem A or therminol 66 and used in the synthesis of high quality CdSe quantum dots (nano crystal made up of semiconductor materials). These quantum dots find application in transistors, diode lasers and solar cells. It is also used as a solvent for the synthesis of iron oxide nanospheres.
Solubility
Miscible with alcohol, acetone and ether. Immiscible with water.
Notes
Avoid reaction with strong Lewis or mineral acids. Free radical initiators should be avoided. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | 15°C to 17°C |
Density | 0.789 |
Boiling Point | 178°C to 179°C (15 mmHg) |
Flash Point | 154°C (309°F) |
Refractive Index | 1.444 |
Quantity | 250 mL |
Beilstein | 1761067 |
Solubility Information | Miscible with alcohol,acetone and ether. Immiscible with water. |
Formula Weight | 252.48 |
Percent Purity | 90% |
Show More |
RUO – Research Use Only