missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, ReagentPlus™, 99%, Honeywell™

Catalog Number p-7120036
Click to view available options
Quantity:
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO
111-87-5
C8H18O
130.23
MFCD00002988
KBPLFHHGFOOTCA-UHFFFAOYSA-N
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
957
CHEBI:16188
octan-1-ol
CCCCCCCCO

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -15°C
Color Colorless
Density 0.827g/mL at 25°C
Boiling Point 196°C
Flash Point 80°C (176°F)
Packaging Glass Bottle
Linear Formula CH3(CH2)7OH
Refractive Index n20/D 1.429(lit.)
Quantity 2.5 L
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity 99%
Grade ReagentPlus™
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'