missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-one, 97%, stab. with 0.1% BHA, Thermo Scientific Chemicals

Catalog Number p-7044574
39.53 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4312-99-6
C8H14O
126.199
MFCD00036558
KLTVSWGXIAYTHO-UHFFFAOYSA-N
1-octen-3-one, vinyl amyl ketone, amyl vinyl ketone, pentyl vinyl ketone, unii-7lt7z4q9xr, 1-octene-3-one, fema no. 3515, 7lt7z4q9xr, n-pentyl vinyl ketone, octen-3-one, 1
61346
oct-1-en-3-one
CCCCCC(=O)C=C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4312-99-6
C8H14O
126.199
MFCD00036558
KLTVSWGXIAYTHO-UHFFFAOYSA-N
1-octen-3-one, vinyl amyl ketone, amyl vinyl ketone, pentyl vinyl ketone, unii-7lt7z4q9xr, 1-octene-3-one, fema no. 3515, 7lt7z4q9xr, n-pentyl vinyl ketone, octen-3-one, 1
61346
oct-1-en-3-one
CCCCCC(=O)C=C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.848
Boiling Point 56°C to 60°C (16 mmHg)
Flash Point 35°C (95°F)
Odor Characteristic
Refractive Index 1.436
Quantity 1 g
UN Number UN1224
Beilstein 1700905
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 1-Octen-3-one, stabilized with 0.1% BHA
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'