Learn More
1-Phenyl-1H-tetrazole-5-thiol, 99%
Description
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.
Solubility
Soluble in 5% ethanol, water (partly), acetone, chloroform, and methanol.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | ∼145°C (decomposition) |
| Flash Point | 138°C (280°F) |
| Odor | Odorless |
| Quantity | 500 g |
| UN Number | UN1325 |
| Beilstein | 139068 |
| Solubility Information | Soluble in 5% ethanol,water (partly),acetone,chloroform,and methanol. |
| IUPAC Name | 1-phenyl-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione |
| Formula Weight | 178.22 |
| Percent Purity | 99% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.