missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in dichloromethane, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.
Solubility
Miscible with dioxane, terahydrofuran, dimethyl formamide, polar and aprotic organic solvents.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.
Solubility
Miscible with dioxane, terahydrofuran, dimethyl formamide, polar and aprotic organic solvents.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
Specifications
Quantity | 250 g |
UN Number | UN2922 |
Beilstein | 5079255 |
Solubility Information | Miscible with dioxane,terahydrofuran,dimethyl formamide,polar and aprotic organic solvents. |
Formula Weight | 318.19 |
Concentration or Composition (by Analyte or Components) | ≥50% w/w soln. in dichloromethane |
Chemical Name or Material | 1-Propylphosphonic acid cyclic anhydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'