missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Thio-b-D-glucopyranose 2,3,4,6-Tetraacetate, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-sulfanyloxan-2-yl]methyl acetate |
| CAS | 19879-84-6 |
| Molecular Formula | C14 H20 O9 S |
| Quantity | 500 mg |
| Synonym | 1-Thio-β-D-glucopyranose Tetraacetate, 1-Thio-β-D-glucose Tetraacetate, 2,3,4,6-Tetra-O-acetyl-1-thio-β-D-glucopyranose, 2,3,4,6-Tetra-O-acetyl-1-thio-β-D-glucopyranoside, 2,3,4,6-Tetra-O-acetyl-1-thio-β-D-glycopyranose, NSC 97032, SKF 83940D, Tetra-O-acetyl-1-thio-β-D-glucopyranose, β-D-Thioglucopyranose 2,3,4,6-Tetraacetate |
| InChI Formula | InChI=1S/C14H20O9S/c1-6(15)19-5-10-11(20-7(2)16)12(21-8(3)17)13(14(24)23-10)22-9(4)18/h10-14,24H,5H2,1-4H3/t10-,11-,12+,13-,14+/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@@H](S)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-sulfanyloxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 364.368 |
| Formula Weight | 364.083 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction