missing translation for 'onlineSavingsMsg'
Learn More

1-Triethylsilyl-4-triethylsilyloxy-1-butyne, 97%, Thermo Scientific Chemicals

Catalog Number 15410368
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
160194-28-5
C16H34OSi2
298.62
MFCD00671376,MFCD09259947
WAQSHASXNKNYSS-UHFFFAOYSA-N
1-triethylsilyl-4-triethylsilyloxy-1-butyne, 1-triethylsilyl-4-triethlylsilyloxy-1-butyne, triethyl 4-triethylsilyl but-3-yn-1-yl oxy silane, triethyl 4-triethylsilyl but-3-inyloxy silane
10935420
CC[Si](CC)(CC)OCCC#C[Si](CC)(CC)CC
This item is not returnable. View return policy
160194-28-5
C16H34OSi2
298.62
MFCD00671376,MFCD09259947
WAQSHASXNKNYSS-UHFFFAOYSA-N
1-triethylsilyl-4-triethylsilyloxy-1-butyne, 1-triethylsilyl-4-triethlylsilyloxy-1-butyne, triethyl 4-triethylsilyl but-3-yn-1-yl oxy silane, triethyl 4-triethylsilyl but-3-inyloxy silane
10935420
CC[Si](CC)(CC)OCCC#C[Si](CC)(CC)CC

1-Triethylsilyl-4-triethylsilyloxy-1-butyne is used as pharmaectical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Triethylsilyl-4-triethylsilyloxy-1-butyne is used as pharmaectical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Linear Formula (CH3CH2)3SiOCH2CH2C≡CSi(CH2CH3)3
IUPAC Name triethyl(4-triethylsilylbut-3-ynoxy)silane
Formula Weight 298.62
Percent Purity 97%
Density 0.834
Chemical Name or Material 1-Triethylsilyl-4-triethylsilyloxy-1-butyne

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'