Learn More
uvex 1 X-Tended Support S1 SRC Safety Shoes
Product Code. p-7236900 Shop All Uvex Products
Click to view available options
Width (Metric):
11 cm
Size:
35
36
37
38
39
40
41
42
43
46
47
48
49
50
51
52
Unit Size:
Each
Product Code. 15300793
Brand: uvex 8511835
Only null left
Add to basket
Add to basket
Uvex 1 safety shoes provide optimal protection and support for the individual movements of the human body. Shoes have been designed based on the latest physiological research and innovative technologies, to reduce work-related stresses on the musculoskeletal system and optimize the wearer's well being.
  • Shoes provide optimal protection and support for the individual movements of the human body
  • Designed based on the latest physiological research and innovative technologies, to reduce work-related stresses on the musculoskeletal system and optimize the wearer's well being
  • The ergonomic construction of the Uvex 1 safety shoes offers the perfect balance between protection and comfort
  • Protection class S1 in accordance with EN ISO 20345:2011 with additional marking for excellent slip resistance (SRC)
  • 100% metal-free Uvex Xenova™ toe cap – compact, anatomically formed, not thermally conductive and with good side stability
  • Made from synthetic materials, making them suitable for people allergic to chrome
  • All sole materials are free of silicones, plasticizers, and other paint wetting impairment substances
  • Medial and lateral heel support for the foot with specially designed side frame
  • Meets the ESD specification with bleeder resistor under 35 megaohms
  • Elastic laces for quick, individual adjustment - normal laces are also provided
  • Suitable for orthopaedic adjustments and insoles from Uvex Medicare
  • Available in a variety of widths and sizes
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Gray
Lace Material Elastic
Liner Material Distance Mesh
Material Microvelour
Size 35
Sole Material Dual Density Polyurethane (PU)
Toe Material Composite
Width (Metric) 11 cm
Gender Unisex
Latex Free Yes
Ankle Style Trainers
Waterproof No
EN ISO 20345 S1
Product Type ESD Safety Shoe