Learn More
Hamilton™ 10 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
This category of needles comes without a hub so they cannot be connected to a Hamilton syringe. The point style and length of the tubing can be specified at the time of order.
Brand:  Hamilton™ 22010-01
Additional Details : Weight : 0.01000kg
Product Code. 12321788
  • 117.00€ / Pack of 6
Estimated Shipment: 29-04-2024
to see stock.
Description
Description
Specification Sheet
Gauge10 gauge
Point Style2, 3, or 4
Needle LengthCustom Needle Length
AutoclavableYes
Sales Quantity6 needles per pack
HubNo Hub
Specifications
Specifications
10 G
Hubless Needles