Learn More
Axygen™ 10 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format
For use with Agilent™/Velocity11 automation systems
Brand:  Axygen™ VT-384-10UL-R-S
Additional Details : Weight : 10.90000kg
 View more versions of this product
Product Code. 12551268
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Maxymum Recovery™ technology for ultra-low retention
  • Available in nonsterile or sterile
Order Info
10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
10 μL
Sterile
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Round
50 Racks/Cs.
Polypropylene
Non-Filtered
Clear
DNase, Rnase Free
Rack
SBS Rack
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST