missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1018, G10180, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, AISI 1018, G10180, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Niobium/Titanium/Tungsten/Sulfur/Oxygen/Arsenic/Cobalt/Nitrogen/Phosphorus/Tin/Aluminium/Vanadium/Molybdenum/Chromium/Copper/Nickel/Carbon/Silicon/Manganese/Calcium/Antimony/Lead/Zirconium |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0001 %/Niobium 0.001 %/Titanium 0.001 %/Tungsten 0.002 %/Sulfur 0.0026 %/Oxygen 0.0052 %/Arsenic 0.007 %/Cobalt 0.008 %/Nitrogen 0.0088 %/Phosphorus 0.009 %/Tin 0.01 %/Aluminium 0.011 %/Vanadium 0.019 %/Molybdenum 0.033 %/Chromium 0.079 %/Copper 0.12 %/Nickel 0.145 %/Carbon 0.169 %/Silicon 0.171 %/Manganese 0.68 %/Calcium (0.0003) %/Antimony <0.001 %/Lead <0.001 %/Zirconium <0.002 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction