missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Phosphor Bronze 5% A, CDA 510, C51000, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Phosphor Bronze 5% A, CDA 510, C51000, 100g
Specifications
Specifications
| Chemical Name or Material | Iron/Manganese/Phosphorus/Tin/Aluminium/Arsenic/Antimony/Silver/Lead/Carbon/Cobalt/Nickel/Silicon |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Iron 0.01 %/Manganese 0.01 %/Phosphorus 0.12 %/Tin 4.6 %/Aluminium (<0.01) %/Arsenic (<0.01) %/Antimony (0.01) %/Silver (0.01) %/Lead (0.011) %/Carbon (0.012) %/Cobalt <0.01 %/Nickel <0.01 %/Silicon <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction