missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Silicon Brass, CDA 875, C87500, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Silicon Brass, CDA 875, C87500, 100g
Specifications
Specifications
| Chemical Name or Material | Manganese/Phosphorus/Nickel/Iron/Silicon/Zinc/Copper/Antimony/Tin/Aluminium/Carbon/Silver/Magnesium/Niobium/Oxygen |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Manganese 0.002 %/Phosphorus 0.006 %/Nickel 0.009 %/Iron 0.076 %/Silicon 3.95 %/Zinc 14.02 %/Copper 81.77 %/Antimony (<0.03) %/Tin (0.006) %/Aluminium <0.01 %/Carbon <0.01 %/Silver <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction