Learn More
1-(5-O-Acetyl-beta-D-ribofuranosyl)-4-amino-1,3,5-triazin-2(1H)-one, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 1-(5-O-Acetyl-beta-D-ribofuranosyl)-4-amino-1,3,5-triazin-2(1H)-one |
| CAS | 1174733-90-4 |
| Molecular Formula | C10 H14 N4 O6 |
| Quantity | 100 mg |
| Synonym | 1,3,5-Triazin-2(1H)-one, 1-(5-O-acetyl-β-D-ribofuranosyl)-4-amino-, 1-(5-O-Acetyl-β-D-ribofuranosyl)-4-amino-1,3,5-triazin-2(1H)-one |
| InChI Formula | InChI=1S/C10H14N4O6/c1-4(15)19-2-5-6(16)7(17)8(20-5)14-3-12-9(11)13-10(14)18/h3,5-8,16-17H,2H2,1H3,(H2,11,13,18)/t5-,6-,7-,8-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=NC(=NC2=O)N |
| IUPAC Name | [(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,3,5-triazin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 286.24 |
| Formula Weight | 286.0913 |