missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxy-2-(2’-carbomethoxy)thiophenylnitrobenzene, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 4-Benzyloxy-2-(2'-carbomethoxy)thiophenylnitrobenzene |
| CAS | 329217-03-0 |
| Molecular Formula | C21 H17 N O5 S |
| Quantity | 100 mg |
| Synonym | 4-Benzyloxy-2-(2-Carbomethoxy)Thiophenylnitrobenzene |
| InChI Formula | InChI=1S/C21H17NO5S/c1-26-21(23)17-9-5-6-10-19(17)28-20-13-16(11-12-18(20)22(24)25)27-14-15-7-3-2-4-8-15/h2-13H,14H2,1H3 |
| SMILES | COC(=O)c1ccccc1Sc2cc(OCc3ccccc3)ccc2[N+](=O)[O-] |
| IUPAC Name | methyl 2-(2-nitro-5-phenylmethoxyphenyl)sulfanylbenzoate |
| Molecular Weight (g/mol) | 395.43 |
| Formula Weight | 395.0827 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction