missing translation for 'onlineSavingsMsg'
Learn More

(4aS,7aS)-Octahydro-1H-pyrrolo[3,4-b]pyridine, Mikromol™

Product Code. 30050273
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30050273

Brand: Mikromol MM0885.150100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(4aS,7aS)-Octahydro-1H-pyrrolo[3,4-b]pyridine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S,S)-2,8-Diazabicyclo[4.3.0]nonane
CAS 151213-40-0
Analyte or Component Names (S,S)-2,8-Diazabicyclo[4.3.0]nonane
Shipping Condition Room Temperature
Molecular Formula C7 H14 N2
Certifications/Compliance ISO 9001
Synonym 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aS,7aS)-, 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aS-cis)-, (1S,6S)-2,8-Diazabicyclo[4.3.0]nonane, (1S,6S)-cis-2,8-Diazabicyclo[4.3.0]nonane, (4AS,7aS)-Octahydro-1H-pyrrole[3,4-b]pyridine, (S,S)-2,8-Diazabicyclo[4.3.0]nonane
InChI Formula InChI=1S/C7H14N2/c1-2-6-4-8-5-7(6)9-3-1/h6-9H,1-5H2/t6-,7+/m0/s1
SMILES C1CN[C@@H]2CNC[C@@H]2C1
IUPAC Name (4aS,7aS)-2,3,4,4a,5,6,7,7a-octahydro-1H-pyrrolo[3,4-b]pyridine
Molecular Weight (g/mol) 126.2
Quantity 100 mg
Formula Weight 126.1157 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.