missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ticlopidine EP Impurity G (as Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Ticlopidine EP Impurity G (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 5-(3-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride |
| CAS | 54943-16-7 |
| Analyte or Component Names | 5-(3-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H14 Cl N S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ticlopidine Hydrochloride Imp. G (EP), Ticlopidine Imp. G (EP), 5-(3-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride, Ticlopidine Hydrochloride Imp. G (EP) as Hydrochloride, Ticlopidine Hydrochloride Impurity G as Hydrochloride, Ticlopidine Impurity G as Hydrochloride |
| InChI Formula | InChI=1S/C14H14ClNS.ClH/c15-13-3-1-2-11(8-13)9-16-6-4-14-12(10-16)5-7-17-14;/h1-3,5,7-8H,4,6,9-10H2;1H |
| SMILES | Cl.Clc1cccc(CN2CCc3sccc3C2)c1 |
| IUPAC Name | 5-[(3-chlorophenyl)methyl]-6,7-dihydro-4H-thieno[3,2-c]pyridine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction