Learn More
5,6-trans-Vitamin D2, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 5,6-trans-Vitamin D2 |
| CAS | 51744-66-2 |
| Molecular Formula | C28H44O |
| Quantity | 100 mg |
| Synonym | 9,10-Secoergosta-5,7,10(19),22-tetraen-3-ol','(3β,5E,7E,22E)- (9CI), (5E,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraen-3β-ol, (5E)-Ergocalciferol, 5,6-trans-Ergocalciferol, 5,6-trans-Vitamin D2 |
| InChI Formula | InChI=1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13+/t20-,22+,25-,26+,27-,28+/m0/s1 |
| SMILES | CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C\3/C[C@@H](O)CCC3=C)\CCC[C@]12C |
| IUPAC Name | (1S,3E)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol |
| Molecular Weight (g/mol) | 396.65 |
| Formula Weight | 396.3392 |