Learn More
Benzyl-d7 Chloride, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Benzyl-d7 Chloride |
| CAS | 59502-05-5 |
| Molecular Formula | C7 D7 Cl |
| Quantity | 100 mg |
| Synonym | Benzene-1,2,3,4,5-d5, 6-(chloromethyl-d2)- (ACI), 6-(Chloromethyl-d2)benzene-1,2,3,4,5-d5 (ACI), Benzene-d5, (chloromethyl-d2)- (9CI), 1-[Chloro(dideuterio)methyl]-2,3,4,5,6-pentadeuteriobenzene, Benzyl chloride-d7, Benzyl-d7 chloride, d7-Benzyl chloride, Perdeuteriobenzyl chloride, Benzyl Chloride-D7 |
| InChI Formula | InChI=1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D,6D2 |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])Cl |
| IUPAC Name | 1-[chloro(dideuterio)methyl]-2,3,4,5,6-pentadeuterio-benzene |
| Molecular Weight (g/mol) | 133.63 |
| Formula Weight | 133.0676 |