missing translation for 'onlineSavingsMsg'
Learn More

Butorphanol N-Oxide, Mikromol™

Product Code. 18053860
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
18053860 100 mg 100mg
30050195 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 18053860

Brand: Mikromol MM1192.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Butorphanol N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butorphanol N-Oxide
CAS 112269-63-3
Analyte or Component Names Butorphanol N-Oxide
Shipping Condition Room Temperature
Molecular Formula C21 H29 N O3
Certifications/Compliance ISO 9001
Synonym Morphinan-3,14-diol, 17-(cyclobutylmethyl)-, 17-oxide (9CI), Butorphanol N-oxide
InChI Formula InChI=1S/C21H29NO3/c23-17-7-6-16-12-19-21(24)9-2-1-8-20(21,18(16)13-17)10-11-22(19,25)14-15-4-3-5-15/h6-7,13,15,19,23-24H,1-5,8-12,14H2/t19-,20+,21-,22?/m1/s1
SMILES Oc1ccc2C[C@@H]3[C@]4(O)CCCC[C@]4(CC[N+]3([O-])CC5CCC5)c2c1
Molecular Weight (g/mol) 343.46
Quantity 100 mg
Formula Weight 343.2147 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.