missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclizine N-Oxide Dihydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Cyclizine N-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Cyclizine N-Oxide Dihydrochloride |
| CAS | 2725355-17-7 |
| Analyte or Component Names | Cyclizine N-Oxide Dihydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C18 H22 N2 O . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Cyclizine N-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C18H22N2O.2ClH/c1-20(21)14-12-19(13-15-20)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;;/h2-11,18H,12-15H2,1H3;2*1H |
| SMILES | Cl.Cl.C[N+]1([O-])CCN(CC1)C(c2ccccc2)c3ccccc3 |
| IUPAC Name | 4-benzhydryl-1-methyl-1-oxidopiperazin-1-ium;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction