Learn More
Doxepin EP Impurity A, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Doxepin EP Impurity A
Specifications
Specifications
| Chemical Name or Material | Dibenzo[b,e]oxepin-11(6H)-one (Doxepinone) |
| CAS | 4504-87-4 |
| Analyte or Component Names | Dibenzo[b,e]oxepin-11(6H)-one (Doxepinone) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H10 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Doxepin Hydrochloride Imp. A (EP), Doxepin Imp. A (EP), Doxepin USP Related Compound A, Doxepin USP RC A, Dibenzo[b,e]oxepin-11(6H)-one, Doxepinone, Doxepin Related Compound A, Doxepin Hydrochloride Impurity A, Doxepin Impurity A |
| InChI Formula | InChI=1S/C14H10O2/c15-14-11-6-2-1-5-10(11)9-16-13-8-4-3-7-12(13)14/h1-8H,9H2 |
| SMILES | O=C1c2ccccc2COc3ccccc13 |
| IUPAC Name | 6H-benzo[c][1]benzoxepin-11-one |
| Show More |