missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-oxo-2-Phenylpropanoate (>90%), TRC

Product Code. 30336613
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30336613

Brand: Toronto Research Chemicals TRCB441355100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3-oxo-2-Phenylpropanoate
CAS 17838-69-6
Molecular Formula C11H12O3
Quantity 100 mg
Synonym Ipratropium Bromide Impurity 2
InChI Formula InChI=1S/C11H12O3/c1-2-14-11(13)10(8-12)9-6-4-3-5-7-9/h3-8,10H,2H2,1H3
SMILES CCOC(=O)C(C=O)c1ccccc1
IUPAC Name ethyl 3-oxo-2-phenylpropanoate
Molecular Weight (g/mol) 192.21
Formula Weight 192.0786
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.