missing translation for 'onlineSavingsMsg'
Learn More

Mesterolone, Mikromol™

Product Code. 18032610
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18032610

Brand: Mikromol MM0495.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mesterolone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mesterolone
CAS 1424-00-6
Analyte or Component Names Mesterolone
Shipping Condition Room Temperature
Molecular Formula C20 H32 O2
Certifications/Compliance ISO 17025
Synonym Androviron, MEST, Mesteranum, Mesterolone, Mestoranum, NSC 75054, Proviron, Provirone 25, SH 723, Testiwop, Androstan-3-one, 17-hydroxy-1-methyl-, (1alpha,5alpha,17beta)-, 5alpha-Androstan-3-one, 17beta-hydroxy-1alpha-methyl- (6CI,7CI,8CI), (1alpha,5alpha,17beta)-17-Hydroxy-1-methylandrostan-3-one, 17beta-Hydroxy-1alpha-methyl-5alpha-androstan-3-one, 1alpha-Methyl-17beta-hydroxy-5alpha-androstan-3-one, 1alpha-Methyl-5alpha-androstan-17beta-ol-3-one, 1alpha-Methyl-5alpha-dihydrotestosterone
InChI Formula InChI=1S/C20H32O2/c1-12-10-14(21)11-13-4-5-15-16-6-7-18(22)19(16,2)9-8-17(15)20(12,13)3/h12-13,15-18,22H,4-11H2,1-3H3/t12-,13-,15-,16-,17-,18-,19-,20-/m0/s1
SMILES C[C@H]1CC(=O)C[C@@H]2CC[C@H]3[C@@H]4CC[C@H](O)[C@@]4(C)CC[C@@H]3[C@@]12C
IUPAC Name (1S,5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-1,10,13-trimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 304.47
Quantity 100 mg
Formula Weight 304.2402 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.