missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone 17-phenylpropionate, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Testosterone 17-phenylpropionate
Specifications
Specifications
| Chemical Name or Material | Testosterone Phenylpropionate |
| CAS | 1255-49-8 |
| Analyte or Component Names | Testosterone Phenylpropionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H36 O3 |
| Synonym | Testosterone, hydrocinnamate (6 CI,7 CI,8 CI), Hydrocinnamic acid, ester with testosterone (8 CI), Androject, Hydrocinnamic acid, testosterone ester, NSC 26643, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | C[C@]12 CC[C@H]3 [C@@H](CCC4=CC(=O)CC[C@]34 C)[C@@H]1 CC[C@@H]2 OC(=O)CCc5ccccc5 |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-phenylpropanoate |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction