missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crackle Test Reference Standard: 1.0 wt% H2O in 10W30 Motor Oil, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Crackle Test Reference Standard: 1.0 wt% H2O in 10W30 Motor Oil
Specifications
Specifications
| Chemical Name or Material | Water |
| CAS | 7732-18-5 |
| Analyte or Component Names | Water |
| Shipping Condition | Room Temperature |
| Molecular Formula | H2 O |
| Concentration | 1% |
| Synonym | Water, Dihydrogen oxide |
| InChI Formula | InChI=1S/H2O/h1H2 |
| SMILES | O |
| IUPAC Name | oxidane |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction