missing translation for 'onlineSavingsMsg'
Learn More

Allura Red AC (Technical Grade), TRC

Product Code. 30314034
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30314034

Brand: Toronto Research Chemicals TRCA54875010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allura Red AC (E 129)
CAS 25956-17-6
Molecular Formula C18 H14 N2 O8 S2 . 2 Na
Quantity 10 g
Synonym 2-Naphthalenesulfonic acid, 6-hydroxy-5-[(2-methoxy-5-methyl-4-sulfophenyl)azo]-, disodium salt (9CI), 2-Naphthalenesulfonic acid, 6-hydroxy-5-[(6-methoxy-4-sulfo-m-tolyl)azo]-, disodium salt (8CI), Allura Red 40, Allura Red AC, Allura red, C.I. 16035, C.I. Food Red 17, E 129, E 129 (dye), FD and C Red No. 40, FD&C Red No. 40, FDC Red 40, FDC Red 40 dye, Fancy Red, Food Red 17, Food Red No. 17, Food Red No. 40, Japan Food Red No. 40, Japan Red 40, Red 40, Red No. 40
InChI Formula InChI=1S/C18H16N2O8S2.2Na/c1-10-7-14(16(28-2)9-17(10)30(25,26)27)19-20-18-13-5-4-12(29(22,23)24)8-11(13)3-6-15(18)21;;/h3-9,21H,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2/b20-19+;;
SMILES [Na+].[Na+].COc1cc(c(C)cc1\N=N\c2c(O)ccc3cc(ccc23)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name disodium;6-hydroxy-5-[(E)-(2-methoxy-5-methyl-4-sulfonatophenyl)diazenyl]naphthalene-2-sulfonate
Molecular Weight (g/mol) 496.42
Formula Weight 495.9987
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.