Learn More
Benzoylpaeoniflorin, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Benzoylpaeoniflorin |
| CAS | 38642-49-8 |
| Molecular Formula | C30H32O12 |
| Quantity | 10 mg |
| Synonym | Benzoylpaeoniflorin, [1aR-(1aα,2β,3aα,5α,5aα,5bα)]-5b-[(Benzoyloxy)methyl]tetrahydro-5-hydroxy-2-methyl-2,5-methano-1H-3,4-dioxacyclobuta[cd]pentalen-1a(2H)-yl, 6-benzoate β-D-Glucopyranoside |
| InChI Formula | InChI=1S/C30H32O12/c1-27-14-29(36)19-12-30(27,28(19,26(41-27)42-29)15-38-24(35)17-10-6-3-7-11-17)40-25-22(33)21(32)20(31)18(39-25)13-37-23(34)16-8-4-2-5-9-16/h2-11,18-22,25-26,31-33,36H,12-15H2,1H3/t18-,19-,20-,21+,22-,25+,26-,27?,28+,29-,30+/m1/s1 |
| SMILES | O[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@@]2([C@@]34COC(C5=CC=CC=C5)=O)C[C@H]3[C@@](O[C@@]4([H])O6)(O)C[C@@]26C)O[C@@H]1COC(C7=CC=CC=C7)=O |
| IUPAC Name | ((2R,3S,4S,5R,6S)-6-(((1aR,1a1S,2R,3aR,5S,5aR)-1a1-((benzoyloxy)methyl)-2-hydroxy-5-methyltetrahydro-1H-3,4-dioxa-2,5-methanocyclobuta[cd]pentalen-5a(5H)-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)methyl benzoate |
| Molecular Weight (g/mol) | 584.57 |
| Formula Weight | 584.19 |