missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,3-dibromopropoxy)tetrabromobisphenol A, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Tetrabromobisphenol A dibromopropyl ether |
| CAS | 21850-44-2 |
| Molecular Formula | C21 H20 Br8 O2 |
| Quantity | 10 mg |
| Synonym | Tetrabromobisphenol A-bis(2,3-dibromopropyl ether), Tetrabromobisphenol A-dibromopropyl ether, Benzene, 1,1'-(1-methylethylidene)bis[3,5-dibromo-4-(2,3-dibromopropoxy)- (9CI, ACI), 1,1'-(1-Methylethylidene)bis[3,5-dibromo-4-(2,3-dibromopropoxy)benzene] (ACI), Propane, 2,2-bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]- (8CI), 1,1'-Isopropylidenebis[3,5-dibromo-4-(2,3-dibromopropoxy)benzene], 1,3-Dibromo-5-[2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl]-2-(2,3-dibromopropoxy)benzene, 2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane, 2,2-Bis[4-(2,3-dibromopropoxy)-3,5-dibromophenyl]propane, 2,2-Bis[4-(2,3-dibromopropyloxy)-3,5-dibromophenyl]propane, 2,2-Bis[[3,5-dibromo-4-(2,3-dibromopropyloxy)]phenyl]propane, 3,3',5,5'-Tetrabromobisphenol A bis(2,3-dibromopropyl) ether, 4,4'-Isopropylidenebis[2,6-dibromo-1-(2,3-dibromopropoxy)benzene], Bis(2,3-dibromopropoxy)tetrabromobisphenol A, Bromkal 66-8, D 5532, E-R 08N, FCP 680, FCP 680G, FG 3100, Fire Cut P 680, Fire Cut P 680G, Fire Guard 3100, Flame Cut 121K, Flame Cut 121R, FR 720, GX 5532, HP 800A, OBE, PE 68, PE 68 (fireproofing agent), Pyroguard SR 720, Pyroguard SR 720N, Saytex HP 800A, Saytex HP 800AG, SR 720, SR 720N, TBBPA-DBPE, Tetrabromobisphenol A 2,3-dibromopropyl ether, Tetrabromobisphenol A bis(2,3-dibromopropyl ether), TexFRon 9001, XZ 6800 |
| InChI Formula | InChI=1S/C21H20Br8O2/c1-21(2,11-3-15(26)19(16(27)4-11)30-9-13(24)7-22)12-5-17(28)20(18(29)6-12)31-10-14(25)8-23/h3-6,13-14H,7-10H2,1-2H3 |
| SMILES | CC(C)(c1cc(Br)c(OCC(Br)CBr)c(Br)c1)c2cc(Br)c(OCC(Br)CBr)c(Br)c2 |
| IUPAC Name | 1,3-dibromo-5-[2-[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propan-2-yl]-2-(2,3-dibromopropoxy)benzene |
| Molecular Weight (g/mol) | 943.61 |
| Formula Weight | 935.493 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction