missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bupropion-d9 Hydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | (±)-Bupropion-D9 Hydrochloride |
| CAS | 1189725-26-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 2H9 H9 Cl N O . Cl H |
| Quantity | 10 mg |
| Synonym | 1-Propanone-3,3,3-d3, 1-(3-chlorophenyl)-2-[[1,1-di(methyl-d3)ethyl-2,2,2-d3]amino]-, hydrochloride (1:1), Bupropion-d9 Hydrochloride, (±)-Bupropion-D9 hydrochloride, Bupropion-D9 hydrochloride (N-tert-butyl D9) |
| InChI Formula | InChI=1S/C13H18ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,15H,1-4H3;1H/i2D3,3D3,4D3; |
| SMILES | Cl.[2H]C([2H])([2H])C(NC(C)C(=O)c1cccc(Cl)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 1-(3-chlorophenyl)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]propan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 285.26 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction