missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Docosanoic-7,7,8,8-d4 Acid, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Docosanoic-7,7,8,8-d4 Acid |
| CAS | 1193721-65-1 |
| Molecular Formula | C22 D4 H40 O2 |
| Quantity | 10 mg |
| Synonym | Docosanoic-7,7,8,8-d4 acid (ACI) |
| InChI Formula | InChI=1S/C22H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H,23,24)/i15D2,16D2 |
| SMILES | [2H]C([2H])(CCCCCCCCCCCCCC)C([2H])([2H])CCCCCC(=O)O |
| IUPAC Name | 7,7,8,8-tetradeuteriodocosanoic acid |
| Molecular Weight (g/mol) | 344.6082 |
| Formula Weight | 344.3592 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction