missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoperazine-d3 Dihydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Trifluoperazine-D3 dihydrochloride |
| CAS | 1432064-02-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21 2H3 H21 F3 N3 S . 2 Cl H |
| Quantity | 10 mg |
| Synonym | Trifluoperazine-D3 Dihydrochloride |
| InChI Formula | InChI=1S/C21H24F3N3S.2ClH/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)28-20-8-7-16(15-18(20)27)21(22,23)24;;/h2-3,5-8,15H,4,9-14H2,1H3;2*1H/i1D3;; |
| SMILES | Cl.Cl.[2H]C([2H])([2H])N1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 10-[3-[4-(trideuteriomethyl)piperazin-1-yl]propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride |
| Molecular Weight (g/mol) | 483.44 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction