Learn More
Medchem Express 10mM * 1 mL KS15
Brand:  Medchem Express HY-115672_1mL
Encompass_Preferred
10mM * 1 mL KS15
Product Code. 18600403
  • 330.75€ / 1mL
 Restricted Item
Estimated Shipment: 27-11-2026