Learn More
Medchem Express 10mM * 1 mL Propoxur
Brand:  Medchem Express HY-B0916_1mL
Encompass_Preferred
10mM * 1 mL Propoxur
Product Code. 18752896
  • 91.80€ / 1mL
 Restricted Item
Estimated Shipment: 02-12-2026