Learn More
Biochrom Quartz Cuvette with Stopper and Black Walls, 400 μL Working Volume
Quartz micro cell with stopper (400 μL working volume). Quartz Cuvette with Stopper and Black Walls, 400 μL Working Volume for use with Ultrospec 7000/8000/9000, Ultrospec 2100 Pro, GeneQuant 1300/100.
Brand: Biochrom 80-2002-99
Additional Details : Weight : 0.10000kg
Description
- Compatible with most spectrophotometers
Specifications
Cuvette | |
400 μL | |
10 mm |
Quartz Cuvette with Stopper and Black Walls | |
Quartz |
For research use only.