Learn More
Viking Optical™ 10X Folding Magnifier
Product Code. p-8011080
Encompass_Preferred
Click to view available options
Diameter (Metric) Lens:
19 mm
Unit Size:
Each
Product Code. 12886325
Brand: Viking Optical™ 5609
Only null left
Add to basket
Add to basket
Pocket doublet type, giving tolerably flat field with minimum chromatic aberration. Lens approximately 19mm diameter. Mounted in metal frame.
Specifications
Diameter (Metric) Lens 19 mm