Learn More
Fisherbrand™ 11mm Crimp Cap Vial Kit, Clear Glass, aluminum Cap, Nat Rubber/PTFE septum, pre-crimped
Product Code. 12900921 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12900921 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12900921
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes 100 vials and 100 caps with pre-assembled septa.
  • ND1
  • Glass
  • Clear
  • 1.5mL
  • Crimp Cap
  • Center hole
  • aluminum
• Natural Rubber/Butyl/TEF• • blue• 45° shore A• ND11• 1.0mmReusable two compartment trays protect vials and closure while keeping matching supplies together
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Material (Liner) Natural Rubber/Butyl/TEF
Color Clear
Diameter (Metric) Outer 11.6 mm
Material Hydrolytic Glass (Class I)
Volume (Metric) 2 mL
No. per Pack 100
Closure Material Aluminum
Closure Type Crimp Cap
Marking Spot No
Height (Metric) 32 mm