Learn More
Axygen™ 1250μL Universal Pipette Tips
Shop All Axygen Scientific Products
Click to view available options
Packaging:
Bulk
Rack
Unit Size:
Pack of 1152
Pack of 3456
Pack of 5000
Description
- Matrix-style tips fits Matrix Impact™ and Impact2™ brand pipettors
- Maxymum Recovery™ tips allows complete retrieval of precious samples
- Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
- Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
- Volume range: 15 to 1250μL
- Available in bulk or racked formats
Specifications
Specifications
| Volume (Metric) | 1250 μL |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Certifications/Compliance | RNase, DNase-free |
| Packaging | Rack |
| For Use With (Equipment) | Thermo Scientific Matrix Impact |
| Material | Polypropylene |