Learn More
Sartorius™ 1289 Grade Refined Pulp/Linters/Ash Qualitative Filter Paper
Brand: Sartorius™ FT-3-207-047
Additional Details : Weight : 0.01800kg
Specifications
Qualitative Filter Paper | |
20μm | |
1289 | |
0.21 mm |
Disc | |
Essentially used for analytical and routine analyzes | |
100 Pack | |
47 mm |