Learn More
Sartorius™ 1289 Grade Refined Pulp/Linters/Ash Qualitative Filter Paper
Brand:  Sartorius™ FT-3-207-047
Additional Details : Weight : 0.01800kg
Product Code. 11719455
  • 11.30€ / Pack of 100
In Stock
to see stock.
Specifications
Qualitative Filter Paper
20μm
1289
0.21 mm
Disc
Essentially used for analytical and routine analyzes
100 Pack
47 mm