Learn More
Linker Industrietechnik™ 150 Dia. x 50mm Height Stainless Steel Test Sieves
Brand: Linker Industrietechnik™ 551.550.222-041
Additional Details : Weight : 0.61000kg
Specifications
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel |
150 mm | |
0.315mm | |
Compliance 2.1 EN 10 204 and ISO 3310-1 | |
50mm |