missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 16-Base (for Autotube) for MPure-32
Shop All MP Biomedicals Products
Click to view available options
Quantity:
300/Pk.
Unit Size:
Pack of 300
Description
- To use with 16-Base on MPure-32 aNAP system.
- Ideal for 1 to 15 runs.
- Each base is designed to install 6 racks of Autotubes.
Specifications
Specifications
| Sample Type | Tissue, Cell Culture, Bacteria |
| For Use With (Equipment) | Autotube |
| Quantity | 300/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction