missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17β-Carboxy-5-oxo-A-nor-3,5-secoandrostan-3-oic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
17β-Carboxy-5-oxo-A-nor-3,5-secoandrostan-3-oic Acid
Specifications
Specifications
| Chemical Name or Material | 17beta-Carboxy-5-oxo-A-nor-3,5-secoandrostan-3-oic Acid |
| CAS | 76763-14-9 |
| Analyte or Component Names | 17beta-Carboxy-5-oxo-A-nor-3,5-secoandrostan-3-oic Acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C19 H28 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Benz[e]indene-6-propanoic acid, 3-carboxydodecahydro-3a,6-dimethyl-7-oxo-, (3S,3aS,5aS,6R,9aS,9bS)-, 1H-Benz[e]indene-6-propanoic acid, 3-carboxydodecahydro-3a,6-dimethyl-7-oxo-, [3S-(3α,3aα,5aβ,6β,9aα,9bβ)]-, (3S,3aS,5aS,6R,9aS,9bS)-3-Carboxydodecahydro-3a,6-dimethyl-7-oxo-1H-benz[e]indene-6-propanoic acid, 17β-Carboxy-5-oxo-A-nor-3,5-secoandrostan-3-oic acid, GR 90434X, Dutasteride Stage 2 related |
| InChI Formula | InChI=1S/C19H28O5/c1-18-9-7-13-11(12(18)4-5-14(18)17(23)24)3-6-15(20)19(13,2)10-8-16(21)22/h11-14H,3-10H2,1-2H3,(H,21,22)(H,23,24)/t11-,12-,13-,14+,18-,19+/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC(=O)[C@]3(C)CCC(=O)O)[C@@H]1CC[C@@H]2C(=O)O |
| IUPAC Name | (3S,3aS,5aS,6R,9aS,9bS)-6-(2-carboxyethyl)-3a,6-dimethyl-7-oxo-1,2,3,4,5,5a,8,9,9a,9b-decahydrocyclopenta[a]naphthalene-3-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction