Learn More
Bochem™ 18/10 E-Poli Crucible Tongs With Ridges
Product Code. 10554261
Change view
Click to view available options
Length (Metric):
160 mm
200 mm
220 mm
250 mm
300 mm
400 mm
500 mm
600 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10554261 600 mm Each
10788941 200 mm Each
10068800 160 mm Each
10232851 220 mm Each
10140001 300 mm Each
10038840 250 mm Each
10282941 400 mm Each
10243041 500 mm Each
8 options
Product Code. 10554261
Brand: Bochem™ 2106
Only null left
Add to basket
Add to basket
With ridges. Bochem™ 18/10 E-Poli Crucible Tongs With Ridges provided secure gripping.
  • With ridges
  • Stainless steel
Specifications
Length (Metric) 600 mm
Material Stainless Steel (18/10)
Type Crucible Tongs