Learn More
Bochem™ 18/10 Stainless Ladle Scoops with Flat Handle
Brand: Bochem™ 8799
Additional Details : Weight : 0.50000kg
Specifications
Ladle Scoop | |
Flat | |
500 mm |
Stainless Steel (18/10) | |
2 L |