Learn More
Bochem™ 18/10 Stainless Steel Dissecting Needle
Click to view available options
For Use With (Equipment):
Type 1 Eye Holder KOLLE 12000
Type 2 Needle Holder KOLLE 12000
Type 3 Lancet Holder KOLLE 12000
Unit Size:
Each
Pack of 10
Diameter (Metric) Inside:
1 mm
1.5 mm
2.5 mm
4 mm
Description
- Type 1: eye
- Type 2: needle
- Type 3: lancet
- For use with Bochem needle holder Kolle 12000
- Manufactured from stainless steel
Specifications
Specifications
| Length (Metric) | 100 mm |
| Material | Stainless Steel (18/10) |
| Product Type | Lancet |
| For Use With (Equipment) | Type 3 Lancet Holder KOLLE 12000 |
| Quantity | 10/pk. |